Learn More
ATCC Enterococcus faecalis; Strain AGR 329
Catalog No. 502383385
Encompass
Enterococcus faecalis strain AGR 329 is a drug-resistant bacterium that was isolated from piglet feces. This whole-genome sequenced strain is resistant to erythromycin and tetracycline; no vancomycin resistance genes were detected by PCR. It has applications for antimicrobial resistance research and drug development.
Catalog No. 50-238-3385 Supplier ATCC Supplier No. BAA2128
Enterococcus faecalis strain AGR 329 is a drug-resistant bacterium that was isolated from piglet feces. This whole-genome sequenced strain is resistant to erythromycin and tetracycline; no vancomycin resistance genes were detected by PCR. It has applications for antimicrobial resistance research and drug development.