Learn More
ATCC Enterococcus faecium
Catalog No. 502383112
Encompass
Enterococcus faecium is a multidrug-resistant, whole-genome sequenced bacterium that was isolated from human feces. This strain is confirmed by PCR to carry the vanA vancomycin resistance gene. It has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-3112 Supplier ATCC Supplier No. BAA2316
Enterococcus faecium is a multidrug-resistant, whole-genome sequenced bacterium that was isolated from human feces. This strain is confirmed by PCR to carry the vanA vancomycin resistance gene. It has applications in antimicrobial resistance research and drug development.