Learn More
ATCC Enterococcus faecium
Supplier:  ATCC BAA2318
Encompass
This whole-genome sequenced strain of Enterococcus faecium was isolated from human feces. This strain is resistant to vancomycin and teicoplanin and was confirmed by PCR to carry the vanA gene. It has applications in antimicrobial resistance research and drug development research.
Catalog No. 50-238-3449