Learn More
ATCC Enterococcus faecium
Catalog No. 502383273
Encompass
Enterococcus faecium strain LRA 55 03 77 [API SA] is a whole-genome sequenced strain that can be used for water testing or as a quality control strain for API products.
Catalog No. 50-238-3273 Supplier ATCC Supplier No. 35667
 Restricted Item
Add to Cart
Add to Cart
Enterococcus faecium strain LRA 55 03 77 [API SA] is a whole-genome sequenced strain that can be used for water testing or as a quality control strain for API products.