Learn More
ATCC Enterococcus faecium
Supplier:  ATCC BAA2317
Encompass
This whole-genome sequenced strain of Enterococcus faecium was isolated from human feces. It is multidrug resistant and was confirmed by PCR to harbor the vanA gene. It has applications in antimicrobial resistance research and drug development.
Catalog No. 50-238-3089