Learn More
ATCC Enterococcus gallinarum
Catalog No. 502382172
Encompass
Enterococcus gallinarum strain NCDO 2311 is a whole-genome sequenced bacterium with applications as a quality control strain. This bacterial culture is typed as Lancefield’s Group D and was confirmed by PCR to have the vanC-1 gene.
Catalog No. 50-238-2172 Supplier ATCC Supplier No. 49608
Enterococcus gallinarum strain NCDO 2311 is a whole-genome sequenced bacterium with applications as a quality control strain. This bacterial culture is typed as Lancefield’s Group D and was confirmed by PCR to have the vanC-1 gene.