Learn More
ATCC Enterococcus gallinarum
Supplier:  ATCC 49609
Encompass
Enterococcus gallinarum strain NCDO 1618 [B74, NCTC 8741] can be used in antimicrobial resistance research and drug development. This strain is typed as serotype 15 and was confirmed by PCR to have the vanC-1 gene.
Catalog No. 50-238-4624