Learn More
ATCC Enterococcus gallinarum; Strain NCDO 2707
Supplier:  ATCC 49610
Encompass
Enterococcus gallinarum strain NCDO 2707 [17R] was isolated from a chicken's duodenum. This whole-genome sequenced bacterial strain was typed as Lancefield’s group D and was confirmed by PCR to have the vanC-1 gene. The bacterium can be used in antimicrobial resistance research and drug development.
Catalog No. 50-238-2759