Learn More
ATCC Enterococcus hirae; Strain FDA M19
Catalog No. 502382827
Encompass
Enterococcus hirae strain FDA M19 is a whole-genome sequenced bacterium with applications in quality control, sterility testing, and bactericide testing.
Catalog No. 50-238-2827 Supplier ATCC Supplier No. 10541
Add to Cart
Add to Cart
Enterococcus hirae strain FDA M19 is a whole-genome sequenced bacterium with applications in quality control, sterility testing, and bactericide testing.