Learn More
Genscript Corporation Antibody Enzyme Proteins Proteins 100 UN 1/EA
Supplier:  Genscript Corporation Z03747100
Encompass
GenScript Enterokinase Animal-Free His Bovine is the light chain of recombinant bovine enterokinase that is a single glycosylated polypeptide chain containing 200 amino acids and is produced under animal free process which is more friendly and safer for drug or vaccine development manufacture and other applications. This product is designed with a 6 × His tag for further removing from the digestion system by being captured by Ni2+ affinity chromatography.
Catalog No. 50-252-9564