Learn More
Genscript Corporation Antibody Enzyme Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0337610
Encompass
Enterokinase (EK) is an enzyme produced by cells of the duodenum and involved in human digestion. It plays a role of turning trypsinogen to its active form trypsin and indirectly activates the pancreatic digestive enzymes. Enterokinase is a specific protease that cleaves after a lysine preceded by four aspartic acids: Asp-Asp-Asp-Asp-Lys(DDDDK↑). Enterokinase will not work if the recognition site is followed by a proline. rbEKhas the highest activity than EK of other species and is used wildly in biochemical applications. rbEK with 6 × His-tag binds with Ni2+ affinity chromatography and was designed to be removed from digestion system. Recombinant Bovine Enterokinase (His-tagged) (rbEK) as the light chain is a single glycosylated polypeptide chain containing 200 amino acids 6 × His at C-terminus. A fully biologically active molecule rbEK has a molecular mass of 40 kDa and is obtained by proprietary chromatographic techniques at GenScript.
Catalog No. 50-252-9645