Learn More
Genscript Corporation Enterokinase, His, Bovine
Catalog No. 502272163
Encompass
Enterokinase (EK) is an enzyme produced by cells of the duodenum and involved in human digestion. It plays a role of turning trypsinogen to its active form trypsin, and indirectly activates the pancreatic digestive enzymes. Enterokinase is a specific protease that cleaves after a lysine preceded by four aspartic acids: Asp-Asp-Asp-Asp-Lys. Enterokinase will not work if the recognition site is followed by a proline. rbEK with 6 × His-tag binds with Ni2+ affinity chromatography and was designed for removing from digestion system.
This item is not returnable. View return policy
Catalog No. 50-227-2163 Supplier Genscript Corporation Supplier No. Z03004100
Enterokinase (EK) is an enzyme produced by cells of the duodenum and involved in human digestion. It plays a role of turning trypsinogen to its active form trypsin, and indirectly activates the pancreatic digestive enzymes. Enterokinase is a specific protease that cleaves after a lysine preceded by four aspartic acids: Asp-Asp-Asp-Asp-Lys. Enterokinase will not work if the recognition site is followed by a proline. rbEK with 6 × His-tag binds with Ni2+ affinity chromatography and was designed for removing from digestion system.