Learn More
New England Biolabs, Inc. Enterokinase, light chain – 2560 units
SDP
Catalog No. 50811909
Encompass
Enterokinase is a specific protease that cleaves after lysine at its cleavage site Asp-Asp-Asp-Asp-Lys. It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate.
Catalog No. 50-811-909 Supplier New England Biolabs, Inc. Supplier No. P8070L
Enterokinase is a specific protease that cleaves after lysine at its cleavage site Asp-Asp-Asp-Asp-Lys. It will sometimes cleave at other basic residues, depending on the conformation of the protein substrate.