Learn More
Medchemexpress LLC Enterolactone | 78473-71-9 | 98.2% | 298.33 | 50 MG
SDP
Catalog No. 5000346117
Encompass_Preferred
Enterolactone is a bioactive phenolic metabolite and mammalian lignan derived from dietary lignans. It possesses estrogenic properties and exhibits anti-breast cancer activity, acting as a radiosensitizer for human breast cancer cell lines by impairing DNA repair and increasing apoptosis.
  • Possesses estrogenic properties.
  • Exhibits anti-breast cancer activity.
  • Acts as a radiosensitizer for human breast cancer cell lines.
  • Arrests the growth of MDA-MB-231 breast cancer cells in the 'S' phase.
  • Triggers apoptosis in MDA-MB-231 breast cancer cells via caspase-3 activation.
  • Inhibits TGF-β-induced migration and invasion of MDA-MB-231 breast cancer cells.
  • Inhibits the TGF-β-induced EMT program in MDA-MB-231 breast cancer cells.
Catalog No. 50-003-46117 Supplier Medchemexpress LLC Supplier No. HY108692500UG
Enterolactone is a bioactive phenolic metabolite and mammalian lignan derived from dietary lignans. It possesses estrogenic properties and exhibits anti-breast cancer activity, acting as a radiosensitizer for human breast cancer cell lines by impairing DNA repair and increasing apoptosis.
  • Possesses estrogenic properties.
  • Exhibits anti-breast cancer activity.
  • Acts as a radiosensitizer for human breast cancer cell lines.
  • Arrests the growth of MDA-MB-231 breast cancer cells in the 'S' phase.
  • Triggers apoptosis in MDA-MB-231 breast cancer cells via caspase-3 activation.
  • Inhibits TGF-β-induced migration and invasion of MDA-MB-231 breast cancer cells.
  • Inhibits the TGF-β-induced EMT program in MDA-MB-231 breast cancer cells.