Learn More
Enzo Life Sciences Enterostatin (human) (1mg). CAS: 117830-79-2
SDP
Supplier:  Enzo Life Sciences BMLP2340001
Encompass_Preferred
This pentapeptide, APGPR, has a number of physiological activities, including inhibition of insulin secretion, suppression of fat intake, and memory enhancement. In vivo, it is generated by the cleavage of its precursor protein, procolipase. Purity: ≥95%. Solubility: Soluble in Water. Long Term Storage: -20°C.
Catalog No. 50-201-1301