Learn More
Cayman Chemical EnterostatInhumnmouser 10mg
SDP
Catalog No. 502947575
Encompass_Preferred
A pentapeptide involved in the regulation of food intake; binds to mitochondrial complex V, also known as F1F0 ATP synthase in a cell-free assay; inhibits ATP production and increases thermogenesis in INS-1 insulinoma cells; reduces body weight in rats fed a high-fat diet; reduces food intake in wild-type, but not Mc4r-/-, mice at 1 nmol, i.c.v.; enhances memory consolidation in mice in a passive avoidance test at 300 mg/kg
Catalog No. 50-294-7575 Supplier Cayman Chemical Supplier No. 3462410 MG
A pentapeptide involved in the regulation of food intake; binds to mitochondrial complex V, also known as F1F0 ATP synthase in a cell-free assay; inhibits ATP production and increases thermogenesis in INS-1 insulinoma cells; reduces body weight in rats fed a high-fat diet; reduces food intake in wild-type, but not Mc4r-/-, mice at 1 nmol, i.c.v.; enhances memory consolidation in mice in a passive avoidance test at 300 mg/kg