Learn More
Bachem Enterotoxin STp (E. coli) Trifluoroacetate
SDP
Supplier:  Bachem 4044297.1000
Encompass_Preferred
Enterotoxin STp (E. coli) Trifluoroacetate, H-Asn-Thr-Phe-Tyr-Cys-Cys-Glu-Leu-Cys-Cys-Asn-Pro-Ala-Cys-Ala-Gly-Cys-Tyr-OH, 115474-04-9, C81H110N20O26S6, Mr 1972.28, 1mg. STp (E. coli), Enterotoxin STp Precursor (55-72) (E. coli). Enterotoxigenic E.coli are able to produce several toxic peptides which may cause acute diarrhea in humans and domestic animals. The E.coli heat-stable enterotoxin STp is syntheszied in vivo as a 72 amino acid precursor consisting of pre-, pro-, and mature region. Mature STp is composed of 18 amino acids containing three intramolecular disulfide bonds, which seem to be important for the correct conformation of the biologically active structure.
Catalog No. 50-260-051