Learn More
Medchemexpress LLC Enterotoxin STp (E. coli) TFA | 98.1% | 1972.25 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYP5089A500UG
Encompass_Preferred
Enterotoxin STp (E. coli) TFA is a heat-stable enterotoxin, suitable for use in vaccine research. It presents as a white to off-white solid.
  • Used in vaccine research
  • Heat-stable enterotoxin
  • White to off-white solid appearance
  • Soluble in DMSO (≥ 100 mg/mL)
  • For research use only
Catalog No. 50-003-98403