Learn More
ATCC Enterovirus A71; Strain: BrCr
Catalog No. 502383351
Encompass
Enterovirus A71 strain BrCr is propagated in Vero cells (ATCC CCL-81). This strain was originally isolated from the stool specimen of a 2-month-old male with aseptic meningitis and was derived by passage in Vero cells. It has applications in enteric disease research.
Catalog No. 50-238-3351 Supplier ATCC Supplier No. VR1775
Enterovirus A71 strain BrCr is propagated in Vero cells (ATCC CCL-81). This strain was originally isolated from the stool specimen of a 2-month-old male with aseptic meningitis and was derived by passage in Vero cells. It has applications in enteric disease research.