Learn More
ATCC Enterovirus D68 RNA; Strain: US/MO/14-18947
Catalog No. 502381972
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from RD cells (ATCC CCL-136™) infected with Enterovirus D68, strain US/MO/14-18947 (ATCC VR-1823™). This product was prepared using methods known to inactivate viruses. Source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-1972 Supplier ATCC Supplier No. VR1823D
Genomic RNA isolated from a preparation of cell lysate and supernatant from RD cells (ATCC CCL-136™) infected with Enterovirus D68, strain US/MO/14-18947 (ATCC VR-1823™). This product was prepared using methods known to inactivate viruses. Source organism and host cells are also available through the ATCC catalog.