Learn More
ATCC Enterovirus D68 ;Strain: US/MO/14-18947
Catalog No. 502385063
Encompass
Enterovirus D68 strain US/MO/14-18947 is propagated in RD cells (ATCC CCL-136). This strain was isolated from a nasopharyngeal swab from a patient with respiratory illness in Missouri in 2014 and was deposited by the Centers for Disease Control and Prevention. It has applications in infectious disease and respiratory disease research.
Catalog No. 50-238-5063 Supplier ATCC Supplier No. VR1823
Enterovirus D68 strain US/MO/14-18947 is propagated in RD cells (ATCC CCL-136). This strain was isolated from a nasopharyngeal swab from a patient with respiratory illness in Missouri in 2014 and was deposited by the Centers for Disease Control and Prevention. It has applications in infectious disease and respiratory disease research.