Learn More
Aviva Systems Biology Corp ENTHD1 Antibody - middle region (ARP53010_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53010_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENTHD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ENTHD1. Alternate names: FLJ25421, dJ370M22.3, CACNA1IApplicationWB. Species reactivity: Human
Catalog No. 50-014-60895