Learn More
Medchemexpress LLC HY-12163 100mg , Entinostat CAS:209783-80-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12163100MG
Encompass_Preferred
HY-12163 100mg Entinostat CAS: 209783-80-2 Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9322