Learn More
TARGETMOL CHEMICALS INC ENTINOSTAT 100MG
Supplier: TARGETMOL CHEMICALS INC T6233100MG
Also available in 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Entinostat (MS-275) (MS-275) is an inhibitor of HDACs that inhibits HDAC1 and HDAC3 (IC50s 0.18/0.74 uM). purity: 99%