Learn More
Apexbio Technology LLC Entinostat (MS-275,SNDX-275) 209783-80-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061949
Encompass_Preferred
Entinostat (MS-275 SNDX-275 CAS 209783-80-2) is an orally bioavailable small molecule inhibitor targeting histone deacetylases (HDACs) a family of enzymes involved in transcriptional regulation by modifying histone acetylation Specifically entinostat exhibits inhibitory activity against class I HDAC enzymes HDAC1 HDAC3 and HDAC8 with reported IC50 values of 0 368 M 0 501 M and 63 4 M respectively In preclinical investigations entinostat demonstrates anti-proliferative effects in diverse human tumor cell lines including breast colon lung multiple myeloma ovarian pancreatic prostate cancers and leukemia Due to these properties entinostat is widely utilized in oncology research
Catalog No. 50-000-61949 Supplier Apexbio Technology LLC Supplier No. A81715.1
Entinostat (MS-275 SNDX-275 CAS 209783-80-2) is an orally bioavailable small molecule inhibitor targeting histone deacetylases (HDACs) a family of enzymes involved in transcriptional regulation by modifying histone acetylation Specifically entinostat exhibits inhibitory activity against class I HDAC enzymes HDAC1 HDAC3 and HDAC8 with reported IC50 values of 0 368 M 0 501 M and 63 4 M respectively In preclinical investigations entinostat demonstrates anti-proliferative effects in diverse human tumor cell lines including breast colon lung multiple myeloma ovarian pancreatic prostate cancers and leukemia Due to these properties entinostat is widely utilized in oncology research