Learn More
Medchemexpress LLC Entinostat-d4 | 209783-80-2 | 380.43 | 500 UG
SDP
Catalog No. 5000437299
Encompass_Preferred
Entinostat-d4 is the deuterium labeled version of Entinostat. Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3. This compound can be used as a tracer and as an internal standard for quantitative analysis. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements, and deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs
  • Intended for research use only
Catalog No. 50-004-37299 Supplier Medchemexpress LLC Supplier No. HY12163S500UG
Entinostat-d4 is the deuterium labeled version of Entinostat. Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3. This compound can be used as a tracer and as an internal standard for quantitative analysis. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements, and deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs
  • Intended for research use only