Learn More
Medchemexpress LLC Entinostat-d4 | 380.43 | 1 MG
SDP
Catalog No. 5000437797
Encompass_Preferred
Entinostat-d4 is the deuterium labeled Entinostat. It is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37797 Supplier Medchemexpress LLC Supplier No. HY12163S1MG
Entinostat-d4 is the deuterium labeled Entinostat. It is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.