Learn More
Selleck Chemical LLC Entinostat (MS-275) 200mg 209783-80-2 SNDX-275
SDP
Supplier:  Selleck Chemical LLC S1053200MG
Encompass_Preferred
Entinostat (MS-275, SNDX-275) strongly inhibits HDAC1 and HDAC3 with IC50 of 0.51 ?M and 1.7 ?M in cell-free assays, compared with HDACs 4, 6, 8, and 10. Entinostat induces autophagy and apoptosis. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0883647