Learn More
Medchemexpress LLC HY-15968 50mg Medchemexpress, Entospletinib CAS:1229208-44-9 Purity:>98%
SDP
Catalog No. 502029183
Encompass_Preferred
Medchemexpress, HY-15968 50mg Entospletinib CAS:1229208-44-9 Entospletinib (GS-9973) is an orally bioavailable, selective Syk inhibitor with an IC50 of 7.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9183 Supplier Medchemexpress LLC Supplier No. HY1596850MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-15968 50mg Entospletinib CAS:1229208-44-9 Entospletinib (GS-9973) is an orally bioavailable, selective Syk inhibitor with an IC50 of 7.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.