Learn More
Aviva Systems Biology Corp ENTPD1 Antibody - N-terminal region (ARP61261_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61261_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENTPD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ENTPD1. Alternate names: ATPDase, CD39, DKFZp686D194, DKFZp686I093, FLJ40921, FLJ40959, NTPDase-1ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-287-314