Learn More
Aviva Systems Biology Corp ENTPD5 Antibody - N-terminal region (ARP62029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENTPD5. It was validated on Western Blot. . Alternate names: CD39L4, MGC163357, MGC163359, NTPDase-5, PCPHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-548