Learn More
Medchemexpress LLC Entrectinib-d8 | 2251773-94-9 | 99.8% | 568.69 | C31H26D8F2N6O2 | 5 MG
SDP
Catalog No. 5000264852
Encompass_Preferred
Entrectinib-d8 is a deuterium-labeled analogue of entrectinib used as an internal standard and reference material for quantitative LC-MS/MS, ADME, and drug metabolism studies. Supplied as a high-purity powder with good solubility in DMSO, it preserves the parent compound's mass-spectrometric behavior while enabling precise tracer-based quantitation. Formula: C31H26D8F2N6O2; molecular weight: 568.69.
  • Deuterium-labeled for reliable mass-spectrometry quantitation.
  • High reported purity for analytical applications.
  • Good solubility in DMSO (≈100 mg/mL).
  • Powder form suitable for long-term storage.
  • Suitable for ADME, metabolite, and tracer studies.
  • Facilitates accurate internal standardization in LC-MS/MS assays.
Catalog No. 50-002-64852 Supplier Medchemexpress LLC Supplier No. HY12678S15MG
Entrectinib-d8 is a deuterium-labeled analogue of entrectinib used as an internal standard and reference material for quantitative LC-MS/MS, ADME, and drug metabolism studies. Supplied as a high-purity powder with good solubility in DMSO, it preserves the parent compound's mass-spectrometric behavior while enabling precise tracer-based quantitation. Formula: C31H26D8F2N6O2; molecular weight: 568.69.
  • Deuterium-labeled for reliable mass-spectrometry quantitation.
  • High reported purity for analytical applications.
  • Good solubility in DMSO (≈100 mg/mL).
  • Powder form suitable for long-term storage.
  • Suitable for ADME, metabolite, and tracer studies.
  • Facilitates accurate internal standardization in LC-MS/MS assays.