Learn More
Selleck Chemical LLC Entrectinib (RXDX-101) 5mg 1108743-60-7 NMS-E628
SDP
Supplier:  Selleck Chemical LLC S79985MG
Encompass_Preferred
Entrectinib (RXDX-101, NMS-E628) is an orally bioavailable pan-TrkA/B/C, ROS1 and ALK inhibitor with IC50 ranging between 0.1 and 1.7 nM. Entrectinib (RXDX-101) induces autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5520