Learn More
Cayman Chemical EnTRopeptIdase FluorogenIc
SDP
Catalog No. 502964347
Encompass_Preferred
A substrate for enteropeptidase; enteropeptidase recognizes amino acid sequence DDDDK and cleaves after the lysine residue, releasing AFC, which displays ex/em spectra of 400/505 nm
Catalog No. 50-296-4347 Supplier Cayman Chemical Supplier No. 22187500 UG
A substrate for enteropeptidase; enteropeptidase recognizes amino acid sequence DDDDK and cleaves after the lysine residue, releasing AFC, which displays ex/em spectra of 400/505 nm