Learn More
ACROBiosystems Envelope protein 50ug
Catalog No. 502535103
Encompass_Preferred
DENV2 (strain Thailand/16681/1984) Envelope Protein His Tag (ENN-D5244) is expressed from human 293 cells (HEK293). It contains AA Met 281 - Ser 677 (Accession # P29990).
Catalog No. 50-253-5103 Supplier ACROBiosystems Supplier No. ENND524450UG
DENV2 (strain Thailand/16681/1984) Envelope Protein His Tag (ENN-D5244) is expressed from human 293 cells (HEK293). It contains AA Met 281 - Ser 677 (Accession # P29990).