Learn More
Med Vet International Sterilization Record Envelope 3M Attest Steam / Eo Gas, 1254E-A, 100/Pack
Catalog No. 502832415
Encompass_Preferred
Contains additional patient information that is suggested for Immediate Use situations Two loads content column Back-up information can be stored neatly inside the envelope There is space for Bowie-Dick test sheets, test pack chemical indicators, temperature/pressure graphs, sterilizer computer printouts, and extra load sheets 100 record envelopes per pack
Catalog No. 50-283-2415 Supplier Med Vet International Supplier No. 471403PK
Contains additional patient information that is suggested for Immediate Use situations Two loads content column Back-up information can be stored neatly inside the envelope There is space for Bowie-Dick test sheets, test pack chemical indicators, temperature/pressure graphs, sterilizer computer printouts, and extra load sheets 100 record envelopes per pack