Learn More
MilliporeSigma™ Supelco™ ENVI™-18 DSK SPE Disk
Catalog No. 111004113 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Outer:
47 mm
90 mm
Quantity:
12/Pk.
24/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Quantity
11-100-4113 47 mm 24/Pk.
11-100-3694 90 mm 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4113 Supplier MilliporeSigma™ Supelco™ Supplier No. 57171
Add to Cart
Add to Cart
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Retention Mechanism: Reversed-phase. Sample Matrix Compatibility: Aqueous solutions (drinking water). The SPE membrane equivalents of ENVI-18 and ENVI-8 packed bed SPE sorbents. Porous glass fiber membranes embedded with C18 or C8 modified silica particles. Provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples. Typical applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides.
TRUSTED_SUSTAINABILITY
Specifications
Column Phase ENVI-18
Column Type SPE
For Use With (Application) Food and Beverages
Product Line ENVI
Type SPE Disk
Quantity 24/Pk.
Diameter (Metric) Outer 47 mm
ENVI is a trademark of Sigma-Aldrich Co. LLC