Learn More
Caron Products And Services Environmental Chamber, 3-Door, 75 cu. ft., Temp & RH
Catalog No. NC2676673
Encompass_Preferred
75 ft3 (2,124 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
Catalog No. NC2676673 Supplier Caron Products And Services Supplier No. 7000751
Add to Cart
Add to Cart
Request a Quote
75 ft3 (2,124 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.