Learn More
Chemglass Life Sciences Enviro-Genie, 120V
SDP
Catalog No. 50143368
Encompass
Enviro-Genie, 120V. The Enviro-Genie allows you to refrigerate, incubate, rotate, rock, stir and shake all in one compact unit. The magnetic stainless steel platform may be easily set to rock or rotate almost any container. Precise temperature control and uniformity - range from 4ºC to 75ºC. Peltier controlled refrigeration and heating provides re-circulated air ensuring temperature consistency and uniformity. The shatterproof viewing window allows for inspection of samples without disturbing chamber environment. A variety of accessories are available to provide maximum application versatility. Supplied with magnetic platform, 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips, non-skid tray and stackable wire rack.
Catalog No. 50-143-368 Supplier Chemglass Life Sciences Supplier No. CLS1398001
Add to Cart
Add to Cart
Request a Quote
Enviro-Genie, 120V. The Enviro-Genie allows you to refrigerate, incubate, rotate, rock, stir and shake all in one compact unit. The magnetic stainless steel platform may be easily set to rock or rotate almost any container. Precise temperature control and uniformity - range from 4ºC to 75ºC. Peltier controlled refrigeration and heating provides re-circulated air ensuring temperature consistency and uniformity. The shatterproof viewing window allows for inspection of samples without disturbing chamber environment. A variety of accessories are available to provide maximum application versatility. Supplied with magnetic platform, 1 clip plate each for 10-13 mm, 15-17mm, 28-30mm tubes, 4 bag mounting strips, non-skid tray and stackable wire rack.