Learn More
Shimadzu Scientific Instruments ENVIRO HI SENS CATALYST
Catalog No. NC2458553
Encompass
High sensitivity platinum catalyst for low level TOC analysis. Comes in a pack of 2 which is required for packing a single combustion tube.
Catalog No. NC2458553 Supplier Shimadzu Scientific Instruments Supplier No. 6300099600
High sensitivity platinum catalyst for low level TOC analysis. Comes in a pack of 2 which is required for packing a single combustion tube.