Learn More
Waters Corp Envirogel GPC Cleanup Column, 15 µm, 19 mm X 150 mm, 100 - 10K, Cyclohexane/Ethyl Acetate , 1/pk
Catalog No. 50780810 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Envirogel High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences such as lipids and natural resins from environmental samples as specified in EPA method 3640A. In the past, the cleanup procedure for environmental samples was performed on low-efficiency GPC columns based on packing particle diameters of 37-75 micron (200-400 mesh) Biobeads S-X resins. The high-efficiency Envirogel Cleanup columns increase the speed of this process while simultaneously reducing solvent consumption. They come packed in your choice of Methylene Chloride or a mixture of Cyclohexane/Ethyl acetate to minimize the need for risky solvent switchover.
Catalog No. 50-780-810 Supplier Waters Corp Supplier No. 186001915
The Envirogel High-Resolution GPC cleanup columns are specifically designed to remove low volatility, high molecular weight interferences such as lipids and natural resins from environmental samples as specified in EPA method 3640A. In the past, the cleanup procedure for environmental samples was performed on low-efficiency GPC columns based on packing particle diameters of 37-75 micron (200-400 mesh) Biobeads S-X resins. The high-efficiency Envirogel Cleanup columns increase the speed of this process while simultaneously reducing solvent consumption. They come packed in your choice of Methylene Chloride or a mixture of Cyclohexane/Ethyl acetate to minimize the need for risky solvent switchover.