Learn More
Waters Corp Waters GPC Column, 15 μm, 19 mm X 300 mm, 50 to 1500
Supplier: Waters Corp WAT036554
The High-Resolution GPC cleanup columns remove low volatility, high molecular weight interferences like lipids and natural resins from environmental samples per EPA method 3640A. These high-efficiency cleanup columns increase process speed, reduce solvent consumption, and are packed in Methylene Chloride or Cyclohexane/Ethyl acetate to minimize solvent switchover.
- Chemistry: GPC
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 pH to 12 pH
- Molecular weight range: 50 to 1500
- Particle shape: Spherical
- Particle size: 15 μm
- Format: Column
- Packing solvent: Methylene Chloride
- System: HPLC
- Technique: GPC
- Inner diameter: 19 mm
- Length: 300 mm
- Units per package: 1 per package
- This product contains hazardous material which requires special freight handling, and additional charges may apply.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.