Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters GPC Guard Column, 15 μm Particle, 4.6 mm ID, 30 mm Length

Catalog No. 50780809 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These high-resolution GPC cleanup columns are designed to remove low volatility, high molecular weight interferences from environmental samples as specified in EPA method 3640A. They offer increased cleanup speed and reduced solvent consumption compared to older methods.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 50 to 1000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Endfitting type: Waters
  • Format: Guard column
  • Packing solvent: Cyclohexane/Ethyl Acetate
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 4.6 mm
  • Length: 30 mm
  • Units per package: 1

Catalog No. 50-780-809 Supplier Waters Corp Supplier No. 186001914
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These high-resolution GPC cleanup columns are designed to remove low volatility, high molecular weight interferences from environmental samples as specified in EPA method 3640A. They offer increased cleanup speed and reduced solvent consumption compared to older methods.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 50 to 1000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Endfitting type: Waters
  • Format: Guard column
  • Packing solvent: Cyclohexane/Ethyl Acetate
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 4.6 mm
  • Length: 30 mm
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.