Learn More
International Enviroguard™ Soft Scrubs™ Disposable Pants
Catalog No. 19472726 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-726 Medium
19-472-729 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-472-726 Supplier International Enviroguard™ Supplier No. FS2062BM
This item is not returnable. View return policy
Ideal undergarments where sterile gowning is used.
Precisely cut for an easy fit
  • Soft, smooth denim blue material
  • Reduce cross-contamination
  • Elastic waist
  • Right rear hip pocket
  • Open ankles
  • Reinforced crotch
Specifications
Size Medium
Gender Unisex
Material SMS
Color Blue
Ankle Style Open
No. of Pockets 1
Disposable Single-use
Sterility Non-sterile
Waist Style Elastic