Learn More
Caron Products And Services Environmental Chamber, 10 cu. ft., Temp & RH
Catalog No. NC2462897
Encompass_Preferred
10 ft3 (283 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This benchtop unit is designed in accordance to ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
Catalog No. NC2462897 Supplier Caron Products And Services Supplier No. 7000101
Add to Cart
Add to Cart
Request a Quote
10 ft3 (283 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This benchtop unit is designed in accordance to ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.