Learn More
Labelmaster Inc Environmentally Hazardous Substance 100mm x 100mm Perm. Vinyl, roll of 500
SDP
Catalog No. NC1382918
Encompass_Preferred
Help shippers and transporters easily identify any liquid or solid material, including its mixtures and solutions that are classified as marine pollutants, with Labelmaster’s Environmentally Hazardous Substance Labels. This dead tree, dead fish marking is part of the Globally Harmonized System (GHS) and was incorporated into the 14th edition of the United Nations Model Regulations as a clear way to communicate how contaminants from solid waste enter the soil and ground water systems. Pressure-sensitive for easy application, these permanent vinyl labels pass the 90-day IMO seawater immersion test, measure 100mm x 100mm and are sold in rolls of 500.
Catalog No. NC1382918 Supplier Labelmaster Inc Supplier No. SL701
Help shippers and transporters easily identify any liquid or solid material, including its mixtures and solutions that are classified as marine pollutants, with Labelmaster’s Environmentally Hazardous Substance Labels. This dead tree, dead fish marking is part of the Globally Harmonized System (GHS) and was incorporated into the 14th edition of the United Nations Model Regulations as a clear way to communicate how contaminants from solid waste enter the soil and ground water systems. Pressure-sensitive for easy application, these permanent vinyl labels pass the 90-day IMO seawater immersion test, measure 100mm x 100mm and are sold in rolls of 500.