Learn More
GENERAL BIOPHYSICS LLC Environmental Chamber for Animal Studies
Catalog No. NC3361701
Encompass
Environmental Chamber for MiceVolume: 8 liters (supports 10-12 mice).Features: Gas recirculation system with O₂ and CO₂ concentration monitoring, CO₂ scrubbing.Dimensions: 12 x 12 inches.
Catalog No. NC3361701 Supplier GENERAL BIOPHYSICS LLC Supplier No. ECM
Add to Cart
Add to Cart
Request a Quote
Environmental Chamber for MiceVolume: 8 liters (supports 10-12 mice).Features: Gas recirculation system with O₂ and CO₂ concentration monitoring, CO₂ scrubbing.Dimensions: 12 x 12 inches.