Learn More
Darwin Chambers Company Single Door Environmental Chamber 21 cu. ft.
SDP
Supplier:  Darwin Chambers Company KB030AA
Encompass_Preferred
The KB030 is a 23.9 cubic feet single door environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Silver-colored, polyester-coated aluminum interior and stainless steel floor with coved corners. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.
Catalog No. 50-189-3920
Add to Cart
Request a Quote