Learn More
Caron Products And Services Environmental Chamber, 25 cu. ft., Temp & RH
Catalog No. NC2261979
Encompass_Preferred
25 ft3 (708 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
This item is not returnable. View return policy
Catalog No. NC2261979 Supplier Caron Products And Services Supplier No. 7000251
Add to Cart
Add to Cart
Request a Quote
25 ft3 (708 L) Environmental Chamber controls temp. (5C to 70C) and relative humidity (20 to 98%). This reach-in unit is designed in accordance with ICH Q1A Guidelines. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.