Learn More
Darwin Chambers Company Single Door Environmental Chamber 31 cu. ft.
SDP
Catalog No. 501893922
Encompass_Preferred
The KB034 is a 32.1 cubic feet extra wide single door environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.
Catalog No. 50-189-3922 Supplier Darwin Chambers Company Supplier No. KB034
Add to Cart
Add to Cart
Request a Quote
The KB034 is a 32.1 cubic feet extra wide single door environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 3 stainless steel shelves. The chamber provides 304 series stainless steel interior and door liner, corrosion-resistant; also a heavy-gauge powder-coated, galvanized steel exterior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.